Gene Transcripts

Gene Symbol
Slc30a5
Entrez Gene ID
294698
Gene Name
solute carrier family 30 (zinc transporter), member 5
Gene Aliases
Chromosome Location
Chr. 2: 30815683 - 30845796 on Build Rnor_6.0
UniGene ID
Rn.16711
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106404.1 NP_001099874.1 4 455

View Full Product Details