Gene Transcripts

Gene Symbol
Vom2r45
Entrez Gene ID
295090
Gene Name
vomeronasal 2 receptor, 45
Gene Aliases
RGD1306812, Vmn2r5
Chromosome Location
Chr. 2: 154670917 - 154701255 on Build Rnor_6.0
UniGene ID
Rn.89385
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008761059.1 XP_008759281.1 3 3161
XM_215580.6 XP_215580.2 3 3143

View Full Product Details