Gene Transcripts

Gene Symbol
Mnd1
Entrez Gene ID
295160
Gene Name
meiotic nuclear divisions 1 homolog (S. cerevisiae)
Gene Aliases
RGD1309007
Chromosome Location
Chr. 2: 183073029 - 183140756 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006232551.2 XP_006232613.1 8, 9 766
XM_008761117.1 XP_008759339.1 8, 9 768
XM_215595.7 XP_215595.5 7, 8 846

View Full Product Details