Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001106469.1 | NP_001099939.1 | 4 | 408 | |
XM_006233264.2 | XP_006233326.1 | 5 | 623 | ||
GenBank mRNA | FQ215285.1 | - | 5 | 548 | |
FQ215331.1 | - | 5 | 577 | ||
FQ215936.1 | - | 5 | 518 | ||
FQ216537.1 | - | 5 | 579 | ||
FQ216608.1 | - | 5 | 596 |