Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012479.3 | NP_036611.2 | 2 | 539 | |
GenBank mRNA | AB024334.1 | - | 2 | 491 | |
AK024230.1 | - | 2 | 503 | ||
AK295029.1 | - | 3 | 418 | ||
BC020963.2 | - | 2 | 513 | 3915246 | |
CR541904.1 | - | 2 | 322 | ||
CR541925.1 | - | 2 | 322 |