Gene Transcripts

Gene Symbol
Lcmt2
Entrez Gene ID
296098
Gene Name
leucine carboxyl methyltransferase 2
Gene Aliases
Chromosome Location
Chr. 3: 113089646 - 113091770 on Build Rnor_6.0
UniGene ID
Rn.17983
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011956.1 NP_001011956.1 1 1897
GenBank mRNA BC083783.1 - 1 1897

View Full Product Details