Gene Transcripts

Gene Symbol
Serinc3
Entrez Gene ID
296350
Gene Name
serine incorporator 3
Gene Aliases
Tde1
Chromosome Location
Chr. 3: 160018643 - 160038078 on Build Rnor_6.0
UniGene ID
Rn.3775
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008312.1 NP_001008313.1 6 765
GenBank mRNA BC085853.1 - 6 765
EU220431.1 - 6 711

View Full Product Details