Gene Transcripts

Gene Symbol
Spopl
Entrez Gene ID
296532
Gene Name
speckle-type POZ protein-like
Gene Aliases
RGD1305847
Chromosome Location
Chr. 3: 401461 - 461651 on Build Rnor_6.0
UniGene ID
Rn.198620
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134447.1 NP_001127919.1 3, 4 375
GenBank mRNA BC167106.1 - 3, 4 375

View Full Product Details