Gene Transcripts

Gene Symbol
Olfml2a
Entrez Gene ID
296708
Gene Name
olfactomedin-like 2A
Gene Aliases
Chromosome Location
Chr. 3: 23223468 - 23259318 on Build Rnor_6.0
UniGene ID
Rn.144357
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106572.1 NP_001100042.1 7 1810
XM_006234074.2 XP_006234136.1 8 2018

View Full Product Details