Gene Transcripts

Gene Symbol
Chchd6
Entrez Gene ID
297436
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 6
Gene Aliases
Chromosome Location
Chr. 4: 121340952 - 121565219 on Build Rnor_6.0
UniGene ID
Rn.6212
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106608.1 NP_001100078.1 6 690
XM_006236846.2 XP_006236908.1 6 697
GenBank mRNA FQ219980.1 - 6 671

View Full Product Details