Gene Transcripts

Gene Symbol
ZNF33A
Entrez Gene ID
7581
Gene Name
zinc finger protein 33A
Gene Aliases
KOX2, KOX31, KOX5, NF11A, ZNF11, ZNF11A, ZNF33, ZZAPK
Chromosome Location
Chr. 10: 38010483 - 38067834 on Build GRCh38
UniGene ID
Hs.435774
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278170.1 NP_001265099.1 4 419
NM_001278171.1 NP_001265100.1 6 598
NM_001278173.1 NP_001265102.1 6 554
NM_001278174.1 NP_001265103.1 6 550
NM_001278175.1 NP_001265104.1 6 539
NM_001278176.1 NP_001265105.1 6 555
NM_001278177.1 NP_001265106.1 5 471
NM_001278178.1 NP_001265107.1 5 480
NM_001278179.1 NP_001265108.1 6 595
NM_001324175.1 NP_001311104.1 6 547
NM_001324176.1 NP_001311105.1 5 477
NM_001324177.1 NP_001311106.1 6 555
NM_001324178.1 NP_001311107.1 4 422
NM_006954.1 NP_008885.1 5 475
NM_006974.2 NP_008905.1 5 472
NR_136709.1 5 475
XM_011519650.2 XP_011517952.1 6 544
XM_011519651.2 XP_011517953.1 6 541
XM_011519653.2 XP_011517955.1 4 357
XM_011519654.2 XP_011517956.1 4 354
XM_017016617.1 XP_016872106.1 5 438
GenBank mRNA AK027057.1 - 5 439
AK292844.1 - 5 456
AK303445.1 - 4 414
AL712670.1 - 6 571
AY184389.1 - 3 294
BC017760.1 - 5 444
BC030511.1 - 1 32
BC126119.1 - 3 347 8991835
BF435065.1 - 5 422
D31763.1 - 5 409
DA276183.1 - 6 507
DA757208.1 - 5 461
DB134021.1 - 6 486
DB234051.1 - 6 519
DB235703.1 - 5 476

View Full Product Details