Gene Transcripts

Gene Symbol
ZNF35
Entrez Gene ID
7584
Gene Name
zinc finger protein 35
Gene Aliases
HF.10, HF10, Zfp105
Chromosome Location
Chr. 3: 44648723 - 44660791 on Build GRCh38
UniGene ID
Hs.288658
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003420.3 NP_003411.3 4 1204
XM_017007129.1 XP_016862618.1 4 1178
XM_017007130.1 XP_016862619.1 4 1199
XM_017007131.1 XP_016862620.1 4 1200
GenBank mRNA AK300581.1 - 3 1058
AK314821.1 - 4 1203
BC013597.1 - 4 1182 4186299
BC015892.1 - 3 1036
BT006962.1 - 3 959
X07289.1 - 4 1172

View Full Product Details