Gene Transcripts

Gene Symbol
Rars2
Entrez Gene ID
297969
Gene Name
arginyl-tRNA synthetase 2, mitochondrial
Gene Aliases
Rarsl
Chromosome Location
Chr. 5: 50075525 - 50127619 on Build Rnor_6.0
UniGene ID
Rn.105643
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106643.2 NP_001100113.1 16 1347
XM_006237986.2 XP_006238048.1 16 1355
GenBank mRNA BC158744.1 - 16 1347

View Full Product Details