Gene Transcripts

Gene Symbol
ZNF69
Entrez Gene ID
7620
Gene Name
zinc finger protein 69
Gene Aliases
Cos5, hZNF3
Chromosome Location
Chr. 19: 11887773 - 11980222 on Build GRCh38
UniGene ID
Hs.565280
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321163.1 NP_001308092.1 5 1077
NM_021915.3 NP_068734.1 5 1035
GenBank mRNA AK123536.1 - 2 1330
BC051309.1 - 5 964 5812091

View Full Product Details