Gene Transcripts

Gene Symbol
ZNF74
Entrez Gene ID
7625
Gene Name
zinc finger protein 74
Gene Aliases
COS52, ZFP520, ZNF520, hZNF7
Chromosome Location
Chr. 22: 20394115 - 20408463 on Build GRCh38
UniGene ID
Hs.517418
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256524.1 NP_001243453.1 3 363
NM_001256525.1 NP_001243454.1 2 263
NM_003426.3 NP_003417.2 2 591
NR_046282.1 2 591
GenBank mRNA AB209663.1 - 2 303
AK291358.1 - 3 289
AK299569.1 - 2 189
BC013395.2 - 2 189 3028001
BC048295.1 - 2 176 5242841
CR456616.1 - 2 130
X71623.1 - 1 26
X92715.1 - 1 1192

View Full Product Details