Gene Transcripts

Gene Symbol
Lonrf3
Entrez Gene ID
298322
Gene Name
LON peptidase N-terminal domain and ring finger 3
Gene Aliases
RGD1565451
Chromosome Location
Chr. X: 122937987 - 122971578 on Build Rnor_6.0
UniGene ID
Rn.95673
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191585.1 NP_001178514.1 1 954
XM_006257447.2 XP_006257509.1 1 1002

View Full Product Details