Gene Transcripts

Gene Symbol
Rhox11
Entrez Gene ID
298346
Gene Name
reproductive homeobox 11
Gene Aliases
Chromosome Location
Chr. X: 124228270 - 124236517 on Build Rnor_6.0
UniGene ID
Rn.153915
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024873.1 NP_001020044.1 1 217
GenBank mRNA DQ058658.1 - 1 217

View Full Product Details