Gene Transcripts

Gene Symbol
Tcea3
Entrez Gene ID
298559
Gene Name
transcription elongation factor A (SII), 3
Gene Aliases
Chromosome Location
Chr. 5: 154598765 - 154634687 on Build Rnor_6.0
UniGene ID
Rn.11547
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001015008.1 NP_001015008.1 6 543
GenBank mRNA BC091180.1 - 6 543

View Full Product Details