Gene Transcripts

Gene Symbol
Padi6
Entrez Gene ID
298595
Gene Name
peptidyl arginine deiminase, type VI
Gene Aliases
RGD1560424
Chromosome Location
Chr. 5: 159241291 - 159256749 on Build Rnor_6.0
UniGene ID
Rn.228213
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191766.1 NP_001178695.1 7 802
XM_008764232.1 XP_008762454.1 7 870

View Full Product Details