Gene Transcripts

Gene Symbol
Slc25a34
Entrez Gene ID
298606
Gene Name
solute carrier family 25, member 34
Gene Aliases
Chromosome Location
Chr. 5: 160176122 - 160179978 on Build Rnor_6.0
UniGene ID
Rn.104973
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013936.1 NP_001013958.1 5 1279
GenBank mRNA BC083723.1 - 5 1279

View Full Product Details