Gene Transcripts

Gene Symbol
Angptl6
Entrez Gene ID
298698
Gene Name
angiopoietin-like 6
Gene Aliases
Chromosome Location
Chr. 8: 21895523 - 21902012 on Build Rnor_6.0
UniGene ID
Rn.162982
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106702.1 NP_001100172.1 4, 5 1030
XM_008765921.1 XP_008764143.1 4, 5 919
GenBank mRNA BC166828.1 - 4, 5 993

View Full Product Details