Gene Transcripts

Gene Symbol
Raver1
Entrez Gene ID
298705
Gene Name
ribonucleoprotein, PTB-binding 1
Gene Aliases
Raver1h
Chromosome Location
Chr. 8: 22092084 - 22111863 on Build Rnor_6.0
UniGene ID
Rn.105739
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013939.1 NP_001013961.1 3 394
GenBank mRNA BC083865.1 - 3 394

View Full Product Details