Gene Transcripts

Gene Symbol
Lamb1
Entrez Gene ID
298941
Gene Name
laminin, beta 1
Gene Aliases
Lamb1-1
Chromosome Location
Chr. 6: 50528798 - 50596593 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750137.3 XP_003750185.1 7 1019
XM_006239996.2 XP_006240058.1 8 888
XM_006239997.2 XP_006240059.1 8 897

View Full Product Details