Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001169108.1 | NP_001162579.1 | 5 | 423 | |
XM_006240154.2 | XP_006240216.1 | 5 | 438 | ||
XM_008764696.1 | XP_008762918.1 | 8 | 1222 | ||
XM_008764697.1 | XP_008762919.1 | 7 | 1032 | ||
XM_008764698.1 | XP_008762920.1 | 8 | 1161 | ||
XM_008764699.1 | XP_008762921.1 | 8 | 1245 | ||
XM_008764700.1 | XP_008762922.1 | 8 | 1131 | ||
XM_008764701.1 | XP_008762923.1 | 8 | 1025 | ||
XM_008764702.1 | XP_008762924.1 | 8 | 1121 |