Gene Transcripts

Gene Symbol
Polr3h
Entrez Gene ID
300088
Gene Name
polymerase (RNA) III (DNA directed) polypeptide H
Gene Aliases
Chromosome Location
Chr. 7: 123146248 - 123156558 on Build Rnor_6.0
UniGene ID
Rn.13037
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127295.2 NP_001120767.1 6 495
GenBank mRNA BC167088.1 - 6 495
FQ220736.1 - 6 529
FQ225628.1 - 6 495
FQ229831.1 - 6 530

View Full Product Details