Gene Transcripts

Gene Symbol
Krt73
Entrez Gene ID
300248
Gene Name
keratin 73, type II
Gene Aliases
Kb36
Chromosome Location
Chr. 7: 143399878 - 143408316 on Build Rnor_6.0
UniGene ID
Rn.143275
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008808.1 NP_001008808.1 5 845
XM_008765703.1 XP_008763925.1 5 885

View Full Product Details