Gene Transcripts

Gene Symbol
Pfdn5
Entrez Gene ID
300257
Gene Name
prefoldin subunit 5
Gene Aliases
Chromosome Location
Chr. 7: 143924662 - 143929400 on Build Rnor_6.0
UniGene ID
Rn.3401
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106794.2 NP_001100264.2 2, 3 216
GenBank mRNA BC169128.1 - 2, 3 182
CF976389.1 - 2, 3 220
CF976393.1 - 2, 3 187

View Full Product Details