Gene Transcripts

Gene Symbol
Cbx5
Entrez Gene ID
300266
Gene Name
chromobox homolog 5
Gene Aliases
Chromosome Location
Chr. 7: 144819727 - 144865014 on Build Rnor_6.0
UniGene ID
Rn.101856
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106797.3 NP_001100267.1 6 679
XM_006242391.2 XP_006242453.1 6 743
XM_006242393.2 XP_006242455.1 5 638
XM_008765707.1 XP_008763929.1 5 652
GenBank mRNA BC166908.1 - 6 680
BC168739.1 - 6 680 6918617

View Full Product Details