Gene Transcripts
- Gene Symbol
- Npm1
- Entrez Gene ID
- 25498
- Gene Name
- nucleophosmin (nucleolar phosphoprotein B23, numatrin)
- Gene Aliases
- B23NP
- Chromosome Location
- Chr. 10: 18080949 - 18091063 on Build Rnor_6.0
- UniGene ID
- Rn.54537
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012992.4 |
NP_037124.1
|
9, 10 |
860 |
|
GenBank mRNA |
BC060579.1 |
- |
9, 10 |
864 |
|
BC088088.1 |
- |
9, 10 |
850 |
7382200 |
FQ213938.1 |
- |
6, 7 |
504 |
|
FQ215037.1 |
- |
6, 7 |
504 |
|
FQ216414.1 |
- |
6, 7 |
504 |
|
FQ220615.1 |
- |
6, 7 |
506 |
|
FQ221347.1 |
- |
6, 7 |
504 |
|
FQ225887.1 |
- |
6, 7 |
520 |
|
FQ227209.1 |
- |
6, 7 |
504 |
|
FQ229117.1 |
- |
6, 7 |
501 |
|
FQ229951.1 |
- |
6, 7 |
499 |
|
FQ230591.1 |
- |
6, 7 |
504 |
|
FQ230655.1 |
- |
6, 7 |
506 |
|
FQ233055.1 |
- |
6, 7 |
504 |
|
FQ234255.1 |
- |
6, 7 |
504 |
|
J03969.1 |
- |
9, 10 |
813 |
|
View Full Product Details