Gene Transcripts

Gene Symbol
Cd3g
Entrez Gene ID
300678
Gene Name
CD3 molecule, gamma
Gene Aliases
Chromosome Location
Chr. 8: 49274553 - 49280926 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077646.2 NP_001071114.1 3 250
XM_006242932.2 XP_006242994.1 3 275
GenBank mRNA S79711.1 - 3 250

View Full Product Details