Gene Transcripts

Gene Symbol
Fam46a
Entrez Gene ID
300870
Gene Name
family with sequence similarity 46, member A
Gene Aliases
RGD1311381
Chromosome Location
Chr. 8: 92935474 - 92942263 on Build Rnor_6.0
UniGene ID
Rn.788
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106844.1 NP_001100314.1 2, 3 858
XM_006243474.2 XP_006243536.1 2, 3 841
GenBank mRNA FQ232419.1 - 1, 2 329

View Full Product Details