Gene Transcripts

Gene Symbol
Dus3l
Entrez Gene ID
301122
Gene Name
dihydrouridine synthase 3-like (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 9: 10289440 - 10294212 on Build Rnor_6.0
UniGene ID
Rn.42187
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034923.1 NP_001030095.1 6 1215
XM_006244314.2 XP_006244376.1 6 1370
GenBank mRNA BC105780.1 - 6 1215

View Full Product Details