Gene Transcripts

Gene Symbol
Vom2r76
Entrez Gene ID
301154
Gene Name
vomeronasal 2 receptor, 76
Gene Aliases
Chromosome Location
Chr. 9: 7643533 - 7667888 on Build Rnor_6.0
UniGene ID
Rn.217862
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099505.1 NP_001092975.1 1, 2 205

View Full Product Details