Gene Transcripts

Gene Symbol
Trem2
Entrez Gene ID
301227
Gene Name
triggering receptor expressed on myeloid cells 2
Gene Aliases
Chromosome Location
Chr. 9: 14611551 - 14618081 on Build Rnor_6.0
UniGene ID
Rn.106975
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106884.1 NP_001100354.1 2 435
XM_006244424.2 XP_006244486.1 2 503
XM_006244425.2 XP_006244487.1 2 500
GenBank mRNA FQ229837.1 - 2 332

View Full Product Details