Gene Transcripts
- Gene Symbol
- EVI5
- Entrez Gene ID
- 7813
- Gene Name
- ecotropic viral integration site 5
- Gene Aliases
- EVI-5, NB4S
- Chromosome Location
- Chr. 1: 92508696 - 92792404 on Build GRCh38
- UniGene ID
- Hs.594434
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001308248.1 |
NP_001295177.1
|
2, 3 |
292 |
|
NM_005665.5 |
NP_005656.4
|
2, 3 |
292 |
|
XM_017002269.1 |
XP_016857758.1
|
2, 3 |
290 |
|
XM_017002270.1 |
XP_016857759.1
|
2, 3 |
601 |
|
XM_017002271.1 |
XP_016857760.1
|
2, 3 |
290 |
|
XM_017002272.1 |
XP_016857761.1
|
2, 3 |
290 |
|
XM_017002273.1 |
XP_016857762.1
|
2, 3 |
516 |
|
XM_017002274.1 |
XP_016857763.1
|
2, 3 |
265 |
|
XM_017002275.1 |
XP_016857764.1
|
2, 3 |
253 |
|
XM_017002276.1 |
XP_016857765.1
|
2, 3 |
514 |
|
XM_017002277.1 |
XP_016857766.1
|
2, 3 |
290 |
|
XM_017002278.1 |
XP_016857767.1
|
2, 3 |
290 |
|
XM_017002280.1 |
XP_016857769.1
|
2, 3 |
514 |
|
XM_017002281.1 |
XP_016857770.1
|
2, 3 |
592 |
|
XM_017002282.1 |
XP_016857771.1
|
2, 3 |
290 |
|
XM_017002283.1 |
XP_016857772.1
|
2, 3 |
290 |
|
XM_017002284.1 |
XP_016857773.1
|
2, 3 |
513 |
|
XM_017002285.1 |
XP_016857774.1
|
2, 3 |
513 |
|
GenBank mRNA |
BP282704.1 |
- |
2, 3 |
292 |
|
CN334472.1 |
- |
2, 3 |
489 |
|
View Full Product Details