Gene Transcripts

Gene Symbol
Il17a
Entrez Gene ID
301289
Gene Name
interleukin 17A
Gene Aliases
CTLA-8, IL-17, IL-17A, Il17
Chromosome Location
Chr. 9: 26841299 - 26844786 on Build Rnor_6.0
UniGene ID
Rn.218513
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106897.1 NP_001100367.1 2 203

View Full Product Details