Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012293.2 | NP_036425.1 | 15 | 2012 | |
XM_005264707.3 | XP_005264764.1 | 14 | 1928 | ||
XM_011510396.1 | XP_011508698.1 | 15 | 1769 | ||
XM_011510397.2 | XP_011508699.1 | 14 | 1857 | ||
GenBank mRNA | AF200348.1 | - | 15 | 1971 | |
BC070119.1 | - | 8 | 2511 | ||
BC098579.1 | - | 15 | 1907 | ||
CN359026.1 | - | 2 | 140 | ||
D86983.1 | - | 15 | 1971 | ||
DA551404.1 | - | 4 | 399 | ||
EF090903.1 | - | 15 | 1971 |