Gene Transcripts

Gene Symbol
Tyw5
Entrez Gene ID
301419
Gene Name
tRNA-yW synthesizing protein 5
Gene Aliases
C2orf60, RGD1311269
Chromosome Location
Chr. 9: 64077841 - 64096265 on Build Rnor_6.0
UniGene ID
Rn.14569
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170473.1 NP_001163944.1 5 659
GenBank mRNA BC158619.1 - 5 640
FM112098.1 - 5 374

View Full Product Details