Gene Transcripts

Gene Symbol
Magebl1
Entrez Gene ID
302438
Gene Name
melanoma antigen, family B-like 1
Gene Aliases
RGD1559523
Chromosome Location
Chr. X: 60939260 - 60942845 on Build Rnor_6.0
UniGene ID
Rn.127613
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128493.1 NP_001121965.1 3 259
XM_006257011.2 XP_006257073.1 3 255
GenBank mRNA BC158628.1 - 3 259

View Full Product Details