Gene Transcripts

Gene Symbol
Hist3h2ba
Entrez Gene ID
303175
Gene Name
histone cluster 3, H2ba
Gene Aliases
Chromosome Location
Chr. 10: 45289741 - 45290370 on Build Rnor_6.0
UniGene ID
Rn.39174
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111127.1 NP_001104597.1 1 71

View Full Product Details