Gene Transcripts

Gene Symbol
Ctc1
Entrez Gene ID
303238
Gene Name
CTS telomere maintenance complex component 1
Gene Aliases
RGD1563106
Chromosome Location
Chr. 10: 55596173 - 55616873 on Build Rnor_6.0
UniGene ID
Rn.203763
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107010.1 NP_001100480.1 8 1615
XM_006246699.2 XP_006246761.1 8 1658
XM_006246700.1 XP_006246762.1 5 838

View Full Product Details