Gene Transcripts

Gene Symbol
Hoxb2
Entrez Gene ID
100361765
Gene Name
homeobox B2
Gene Aliases
Hoxbes2
Chromosome Location
Chr. 10: 84200859 - 84203065 on Build Rnor_6.0
UniGene ID
Rn.231109
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047091.1 NP_001040556.1 303
GenBank mRNA DQ399532.1 - 303

View Full Product Details