Gene Transcripts

Gene Symbol
Osbpl7
Entrez Gene ID
303497
Gene Name
oxysterol binding protein-like 7
Gene Aliases
Chromosome Location
Chr. 10: 84986335 - 85003947 on Build Rnor_6.0
UniGene ID
Rn.205144
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107044.1 NP_001100514.1 4, 5 530
XM_006247299.2 XP_006247361.1 5, 6 587
XM_006247300.2 XP_006247362.1 5, 6 527

View Full Product Details