Gene Transcripts

Gene Symbol
Igll1
Entrez Gene ID
100360919
Gene Name
immunoglobulin lambda-like polypeptide 1
Gene Aliases
Chromosome Location
Chr. 11: 88388368 - 88391524 on Build Rnor_6.0
UniGene ID
Rn.110118
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190341.1 NP_001177270.1 3 553
XM_006248656.2 XP_006248718.1 3 525

View Full Product Details