Gene Transcripts

Gene Symbol
Tssk2
Entrez Gene ID
304181
Gene Name
testis-specific serine kinase 2
Gene Aliases
Stk22b
Chromosome Location
Chr. 11: 87235058 - 87236413 on Build Rnor_6.0
UniGene ID
Rn.163190
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012019.1 NP_001012019.1 1 195
GenBank mRNA BC079037.1 - 1 195

View Full Product Details