Gene Transcripts

Gene Symbol
Zan
Entrez Gene ID
304379
Gene Name
zonadhesin
Gene Aliases
Chromosome Location
Chr. 12: 22285377 - 22389008 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008769180.1 XP_008767402.1 6 1448

View Full Product Details