Gene Transcripts

Gene Symbol
Tchp
Entrez Gene ID
304547
Gene Name
trichoplein, keratin filament binding
Gene Aliases
RGD1310813
Chromosome Location
Chr. 12: 47638278 - 47653538 on Build Rnor_6.0
UniGene ID
Rn.19337
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191666.1 NP_001178595.1 6 763
XM_006249451.1 XP_006249513.1 7 789
XM_006249452.1 XP_006249514.1 6 776

View Full Product Details