Gene Transcripts

Gene Symbol
Myo18b
Entrez Gene ID
304551
Gene Name
myosin XVIIIb
Gene Aliases
Chromosome Location
Chr. 12: 49761099 - 49979745 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006249577.2 XP_006249639.1 15 3218
XM_006249578.2 XP_006249640.1 14 3007

View Full Product Details