Gene Transcripts

Gene Symbol
Tfcp2l1
Entrez Gene ID
304741
Gene Name
transcription factor CP2-like 1
Gene Aliases
Tcfcp2l1
Chromosome Location
Chr. 13: 34610689 - 34671617 on Build Rnor_6.0
UniGene ID
Rn.233953
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107170.1 NP_001100640.1 2, 3 305

View Full Product Details