Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001134867.1 | NP_001128339.1 | 3 | 591 | |
XM_006249690.2 | XP_006249752.1 | 3 | 951 | ||
XM_006249694.2 | XP_006249756.1 | 3 | 588 | ||
XM_008769459.1 | XP_008767681.1 | 3 | 588 | ||
XM_008769460.1 | XP_008767682.1 | 3 | 588 | ||
XM_008769461.1 | XP_008767683.1 | 3 | 588 | ||
XM_008769462.1 | XP_008767684.1 | 3 | 588 | ||
XM_008769463.1 | XP_008767685.1 | 3 | 587 | ||
GenBank mRNA | BC087715.1 | - | 2 | 269 |